Gene Transcripts

Gene Symbol
FOXP2
Entrez Gene ID
93986
Gene Name
forkhead box P2
Gene Aliases
CAGH44, SPCH1, TNRC10
Chromosome Location
Chr. 7: 114086310 - 114693772 on Build GRCh38
UniGene ID
Hs.282787
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172766.2 NP_001166237.1 13 1945
NM_014491.3 NP_055306.1 13 1948
NM_148898.3 NP_683696.2 14 2023
NM_148900.3 NP_683698.2 14 1999
NR_033766.1 16 1959
NR_033767.1 14 2006
XM_017012801.1 XP_016868290.1 16 2629
GenBank mRNA AF086040.1 - 3 338
AF337817.1 - 13 1724
AK131266.1 - 16 1943
AK296957.1 - 14 1879
AY144615.1 - 14 2018
BC126104.1 - 14 1680 8991820
BC143866.1 - 14 1680 9052382
BC143867.1 - 13 1619 9052383
CR749236.1 - 20 2329
DQ778626.1 - 8 1019
KJ534839.1 - 13 1649
KJ535079.1 - 12 1787
KJ535086.1 - 12 1573

View Full Product Details