Gene Transcripts

Gene Symbol
OSBPL11
Entrez Gene ID
114885
Gene Name
oxysterol binding protein like 11
Gene Aliases
ORP-11, ORP11, OSBP12, TCCCIA00292
Chromosome Location
Chr. 3: 125528858 - 125595537 on Build GRCh38
UniGene ID
Hs.477440
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022776.4 NP_073613.2 12 2847
GenBank mRNA AF392454.1 - 12 2418
AK023074.1 - 7 1244
AK023226.1 - 7 1277
AK292702.1 - 12 2199
BC010422.1 - 7 1431
BC065213.1 - 12 2197 3916115

View Full Product Details