Gene Transcripts

Gene Symbol
TTC8
Entrez Gene ID
123016
Gene Name
tetratricopeptide repeat domain 8
Gene Aliases
BBS8, RP51
Chromosome Location
Chr. 14: 88824153 - 88877996 on Build GRCh38
UniGene ID
Hs.303055
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001288781.1 NP_001275710.1 11 1067
NM_001288782.1 NP_001275711.1 9 987
NM_001288783.1 NP_001275712.1 10 959
NM_144596.3 NP_653197.2 11 1019
NM_198309.3 NP_938051.1 10 989
NM_198310.3 NP_938052.1 9 899
XM_006720035.1 XP_006720098.1 9 1271
XM_006720037.2 XP_006720100.1 8 1181
XM_011536432.1 XP_011534734.1 10 1349
XM_011536433.1 XP_011534735.1 10 1349
XM_011536434.1 XP_011534736.1 9 1259
XM_011536435.2 XP_011534737.1 9 735
XM_017020977.1 XP_016876466.1 9 1271
XM_017020978.1 XP_016876467.1 8 1181
GenBank mRNA AK093891.1 - 9 837
AK124675.1 - 10 869
AK315772.1 - 9 874
AL833901.1 - 8 723
AY366523.1 - 9 845
AY366524.1 - 8 755
AY373972.1 - 9 845
BC001563.1 - 5 337
BC026351.1 - 10 923 4799330
BC095433.1 - 9 827 30368216
BX161472.1 - 8 827
BX248248.1 - 6 431
KU178849.1 - 10 871
KU178850.1 - 9 793
KU178851.1 - 8 703
KU178852.1 - 4 313

View Full Product Details