Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304499.1 | NP_001291428.1 | 5 | 478 | |
NM_001304500.1 | NP_001291429.1 | 3 | 208 | ||
NM_144600.3 | NP_653201.1 | 4 | 406 | ||
NR_130755.1 | 3 | 321 | |||
NR_130756.1 | 5 | 506 | |||
NR_130757.1 | 2 | 123 | |||
GenBank mRNA | AK055715.1 | - | 4 | 349 | |
AL832498.1 | - | 4 | 369 | ||
AY507846.1 | - | 4 | 362 | ||
BC022321.1 | - | 4 | 357 | 4702249 | |
BI460301.1 | - | 5 | 444 | ||
BI550657.1 | - | 3 | 297 | ||
BU661163.1 | - | 2 | 74 | ||
CB991926.1 | - | 5 | 495 | ||
DA178808.1 | - | 5 | 450 | ||
DB103294.1 | - | 3 | 152 | ||
HY004717.1 | - | 4 | 407 |