Gene Transcripts

Gene Symbol
ANKRD13B
Entrez Gene ID
124930
Gene Name
ankyrin repeat domain 13B
Gene Aliases
Chromosome Location
Chr. 17: 29589769 - 29614761 on Build GRCh38
UniGene ID
Hs.662164
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152345.4 NP_689558.4 11, 12 1360
XM_005257920.4 XP_005257977.1 11, 12 1326
XM_011524319.2 XP_011522621.1 11, 12 1563
XM_017024174.1 XP_016879663.1 11, 12 1260
XM_017024175.1 XP_016879664.1 11, 12 1262
XM_017024176.1 XP_016879665.1 11, 12 1192
GenBank mRNA AK092673.1 - 11, 12 1177
AK097716.1 - 10, 11 1196
AK098421.1 - 3, 4 316
BC032554.1 - 11, 12 1407 5557374
BC062597.1 - 10, 11 1210
BG029526.1 - 141

View Full Product Details