Gene Transcripts

Gene Symbol
OR2T10
Entrez Gene ID
127069
Gene Name
olfactory receptor family 2 subfamily T member 10
Gene Aliases
OR1-64
Chromosome Location
Chr. 1: 248483404 - 248645469 on Build GRCh38
UniGene ID
Hs.553585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004693.1 NP_001004693.1 1 646
XM_017000273.1 XP_016855762.1 2 897
GenBank mRNA BC136937.1 - 1 677
BC136940.1 - 1 677 9020932

View Full Product Details