Gene Transcripts

Gene Symbol
DOCK11
Entrez Gene ID
139818
Gene Name
dedicator of cytokinesis 11
Gene Aliases
ACG, ZIZ2, bB128O4.1
Chromosome Location
Chr. X: 118495664 - 118686162 on Build GRCh38
UniGene ID
Hs.368203
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144658.3 NP_653259.3 23 2578
XM_005262368.3 XP_005262425.1 23 2823
XM_005262369.4 XP_005262426.1 23 2517
XM_011531276.2 XP_011529578.1 23 2458
XM_011531278.2 XP_011529580.1 10 944
GenBank mRNA AK124735.1 - 17 1963
AK125641.1 - 2 90
AY692226.1 - 23 2578

View Full Product Details