Gene Transcripts

Gene Symbol
WIPF2
Entrez Gene ID
147179
Gene Name
WAS/WASL interacting protein family member 2
Gene Aliases
WICH, WIRE
Chromosome Location
Chr. 17: 40219304 - 40284136 on Build GRCh38
UniGene ID
Hs.421622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133264.4 NP_573571.1 5 1159
XM_005257083.2 XP_005257140.1 5 1161
XM_005257084.2 XP_005257141.1 5 1119
XM_011524412.1 XP_011522714.1 4 1060
XM_011524413.1 XP_011522715.1 4 1041
GenBank mRNA AB043786.1 - 4 919
AF370396.1 - 5 1083
AJ431177.1 - 4 919
AK026913.1 - 3 751
AK289578.1 - 5 1119
AK289779.1 - 5 1178
AK290301.1 - 5 1155
AK311544.1 - 4 1583
AK315836.1 - 5 1139
AL834239.1 - 4 1028
BC025965.1 - 5 1115
BC065551.1 - 5 1096 6044222

View Full Product Details