Gene Transcripts

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
Chromosome Location
Chr. 1: 235440654 - 235504481 on Build GRCh38
UniGene ID
Hs.498143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277155.2 NP_001264084.1 5 783
NM_152490.4 NP_689703.1 4 660
XM_006711749.3 XP_006711812.1 4 602
XM_017000394.1 XP_016855883.1 5 783
XM_017000395.1 XP_016855884.1 5 736
GenBank mRNA AB209046.1 - 3 214
AK091497.1 - 3 332
AK130297.1 - 3 503
BC016974.1 - 5 724
BC029564.1 - 4 660 4826983

View Full Product Details