Gene Transcripts

Gene Symbol
SPICE1
Entrez Gene ID
152185
Gene Name
spindle and centriole associated protein 1
Gene Aliases
CCDC52, SPICE
Chromosome Location
Chr. 3: 113442718 - 113515187 on Build GRCh38
UniGene ID
Hs.477144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144718.3 NP_653319.1 7 880
XM_011512461.2 XP_011510763.1 7 637
XM_017005775.1 XP_016861264.1 7 637
XM_017005776.1 XP_016861265.1 7 774
XM_017005777.1 XP_016861266.1 7 697
XM_017005778.1 XP_016861267.1 7 881
GenBank mRNA AK126897.1 - 3 286
AL832148.1 - 7 849
AL832327.2 - 7 663
AY099107.1 - 7 800
BC036951.1 - 7 821 5500345

View Full Product Details