Gene Transcripts

Gene Symbol
NOP9
Entrez Gene ID
161424
Gene Name
NOP9 nucleolar protein
Gene Aliases
C14orf21
Chromosome Location
Chr. 14: 24299854 - 24309126 on Build GRCh38
UniGene ID
Hs.708040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286367.1 NP_001273296.1 5 1205
NM_174913.2 NP_777573.1 5 1205
XM_005267385.1 XP_005267442.1 5 1232
XM_011536526.1 XP_011534828.1 5 1024
XM_011536527.2 XP_011534829.1 5 1232
GenBank mRNA AB095941.1 - 5 1197
BC025332.1 - 5 1183 4549328
BX248018.1 - 5 1167
BX647311.1 - 5 1172

View Full Product Details