Gene Transcripts

Gene Symbol
WBP2NL
Entrez Gene ID
164684
Gene Name
WBP2 N-terminal like
Gene Aliases
GRAMD7, PAWP
Chromosome Location
Chr. 22: 41998725 - 42058456 on Build GRCh38
UniGene ID
Hs.655541
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152613.2 NP_689826.2 3 392
GenBank mRNA AF393575.1 - 1 76
AK128138.1 - 3 392
AK129656.1 - 3 345
BC022546.1 - 3 329 4811804
BC022549.1 - 3 329 4815849
CR456400.1 - 3 305
DB079211.1 - 3 392

View Full Product Details