Gene Transcripts

Gene Symbol
BMPER
Entrez Gene ID
168667
Gene Name
BMP binding endothelial regulator
Gene Aliases
CRIM3, CV-2, CV2
Chromosome Location
Chr. 7: 33904911 - 34155872 on Build GRCh38
UniGene ID
Hs.660998
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133468.4 NP_597725.1 7 916
XM_005249633.2 XP_005249690.1 6 656
XM_017011800.1 XP_016867289.1 6 576
XM_017011801.1 XP_016867290.1 6 576
GenBank mRNA AB075845.1 - 3 183
AK289963.1 - 6 656
AY324650.1 - 7 915
BC060868.1 - 6 593 30345290

View Full Product Details