Gene Transcripts
- Gene Symbol
- DCDC5
- Entrez Gene ID
- 100506627
- Gene Name
- doublecortin domain containing 5
- Gene Aliases
- Chromosome Location
- Chr. 11: 30863603 - 30992686 on Build GRCh38
- UniGene ID
- Hs.530438
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020869.3 |
NP_065920.2
|
9, 10 |
1374 |
|
GenBank mRNA |
BX647348.1 |
- |
9, 10 |
1374 |
|
View Full Product Details