Gene Transcripts

Gene Symbol
ZFC3H1
Entrez Gene ID
196441
Gene Name
zinc finger C3H1-type containing
Gene Aliases
CCDC131, CSRC2, PSRC2
Chromosome Location
Chr. 12: 71609599 - 71663969 on Build GRCh38
UniGene ID
Hs.245798
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144982.4 NP_659419.3 27 5448
GenBank mRNA AB011118.2 - 27 5298
AK125035.1 - 18 2902
BX647709.1 - 27 5328

View Full Product Details