Gene Transcripts

Gene Symbol
ARID2
Entrez Gene ID
196528
Gene Name
AT-rich interaction domain 2
Gene Aliases
BAF200, p200
Chromosome Location
Chr. 12: 45729683 - 45908037 on Build GRCh38
UniGene ID
Hs.317304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152641.2 NP_689854.2 15 2803
XM_006719272.3 XP_006719335.1 15 2957
XM_011538025.1 XP_011536327.1 9 1910
GenBank mRNA AL832200.1 - 11 2356
AY727870.2 - 15 2803
BX537879.1 - 1 431
BX648152.1 - 9 2091
BX649133.1 - 7 1649
CR749833.1 - 14 2722
DQ096628.1 - 15 2803

View Full Product Details