Gene Transcripts

Gene Symbol
ARID2
Entrez Gene ID
196528
Gene Name
AT-rich interaction domain 2
Gene Aliases
BAF200, p200
Chromosome Location
Chr. 12: 45729683 - 45908037 on Build GRCh38
UniGene ID
Hs.317304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152641.2 NP_689854.2 15 2902
XM_006719272.3 XP_006719335.1 15 3056
XM_011538025.1 XP_011536327.1 9 2009
GenBank mRNA AK027718.1 - 1 56
AL832200.1 - 11 2455
AY727870.2 - 15 2902
BX537879.1 - 1 530
BX648152.1 - 9 2190
BX649133.1 - 7 1748
CR749833.1 - 14 2821
DQ096628.1 - 15 2902

View Full Product Details