Gene Transcripts

Gene Symbol
UBR1
Entrez Gene ID
197131
Gene Name
ubiquitin protein ligase E3 component n-recognin 1
Gene Aliases
JBS
Chromosome Location
Chr. 15: 42942897 - 43106088 on Build GRCh38
UniGene ID
Hs.591121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174916.2 NP_777576.1 12 1376
GenBank mRNA AF525401.1 - 12 1259
AK027803.1 - 9 817
AK056441.1 - 12 1326
AK292516.1 - 12 1323
AK302489.1 - 12 1403
AY061886.1 - 12 1310
BC113505.1 - 12 1388 8322557
BC113507.1 - 12 1388 8322559

View Full Product Details