Gene Transcripts

Gene Symbol
SNX32
Entrez Gene ID
254122
Gene Name
sorting nexin 32
Gene Aliases
SNX6B
Chromosome Location
Chr. 11: 65833636 - 65854687 on Build GRCh38
UniGene ID
Hs.591950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152760.2 NP_689973.2 6 668
XM_006718488.3 XP_006718551.1 6 971
XM_017017470.1 XP_016872959.1 6 946
XM_017017471.1 XP_016872960.1 7 1130
XM_017017472.1 XP_016872961.1 5 1332
XM_017017473.1 XP_016872962.1 5 1332
GenBank mRNA AK055496.1 - 6 628
AK289587.1 - 6 634
AK299512.1 - 5 738
AK309789.1 - 4 884
AL833261.1 - 6 654
BC040981.2 - 6 644 4797976
BC045563.1 - 6 617 4815203

View Full Product Details