Gene Transcripts

Gene Symbol
WFDC11
Entrez Gene ID
259239
Gene Name
WAP four-disulfide core domain 11
Gene Aliases
WAP11
Chromosome Location
Chr. 20: 45648563 - 45670239 on Build GRCh38
UniGene ID
Hs.374924
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147197.2 NP_671730.1 3 225
GenBank mRNA AY047609.1 - 3 225
BC062670.1 - 3 256 6101449

View Full Product Details