Gene Transcripts

Gene Symbol
KANSL1
Entrez Gene ID
284058
Gene Name
KAT8 regulatory NSL complex subunit 1
Gene Aliases
CENP-36, KDVS, KIAA1267, MSL1v1, NSL1, hMSL1v1
Chromosome Location
Chr. 17: 46029916 - 46225374 on Build GRCh38
UniGene ID
Hs.648744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193465.1 NP_001180394.1 3 1289
NM_001193466.1 NP_001180395.1 2 1503
NM_015443.3 NP_056258.1 2 1205
XM_006721823.1 XP_006721886.1 3 1289
XM_006721824.3 XP_006721887.1 3 1473
XM_011524628.2 XP_011522930.1 3 1473
XM_011524629.2 XP_011522931.1 3 1473
XM_011524630.2 XP_011522932.1 3 1473
XM_011524631.2 XP_011522933.1 3 1474
XM_017024488.1 XP_016879977.1 3 1289
XM_017024489.1 XP_016879978.1 2 1400
GenBank mRNA AB033093.1 - 1 1126
AK291259.1 - 2 1266
BC098376.1 - 2 1205 6181132
BX538006.1 - 2 1506

View Full Product Details