Gene Transcripts
- Gene Symbol
- RSPO1
- Entrez Gene ID
- 284654
- Gene Name
- R-spondin 1
- Gene Aliases
- CRISTIN3, RSPO
- Chromosome Location
- Chr. 1: 37611350 - 37634923 on Build GRCh38
- UniGene ID
- Hs.135015
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001038633.3 |
NP_001033722.1
|
5, 6 |
1167 |
|
NM_001242908.1 |
NP_001229837.1
|
4, 5 |
988 |
|
NM_001242909.1 |
NP_001229838.1
|
4, 5 |
667 |
|
NM_001242910.1 |
NP_001229839.1
|
4, 5 |
988 |
|
XM_006710583.3 |
XP_006710646.1
|
3, 4 |
903 |
|
GenBank mRNA |
AK098225.1 |
- |
4, 5 |
409 |
|
BC114966.1 |
- |
3, 4 |
585 |
40084277 |
DQ165084.1 |
- |
4, 5 |
730 |
|
DQ165085.1 |
- |
4, 5 |
730 |
|
DQ318235.1 |
- |
5, 6 |
877 |
|
View Full Product Details