Gene Transcripts

Gene Symbol
RGMB
Entrez Gene ID
285704
Gene Name
repulsive guidance molecule family member b
Gene Aliases
DRAGON
Chromosome Location
Chr. 5: 98768386 - 98796494 on Build GRCh38
UniGene ID
Hs.526902
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012761.2 NP_001012779.2 4 1089
XM_005271966.3 XP_005272023.1 4 945
XM_011543345.2 XP_011541647.1 4 850
XM_011543346.2 XP_011541648.1 2 1227
XM_011543347.2 XP_011541649.2 3 773
XM_017009390.1 XP_016864879.1 4 1337
XM_017009391.1 XP_016864880.1 4 1235
GenBank mRNA AK074887.1 - 4 1336
BC067736.1 - 4 1089 30340500

View Full Product Details