Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175858.2 | NP_787054.1 | 1 | 348 | |
GenBank mRNA | AB096963.1 | - | 1 | 326 | |
AJ457065.1 | - | 1 | 348 | ||
BC130555.1 | - | 1 | 341 | 40146586 | |
BC130557.1 | - | 1 | 341 | 40146588 |