Gene Transcripts

Gene Symbol
DCDC1
Entrez Gene ID
341019
Gene Name
doublecortin domain containing 1
Gene Aliases
Chromosome Location
Chr. 11: 31262624 - 31369810 on Build GRCh38
UniGene ID
Hs.127405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181807.3 NP_861523.2 4, 5 682
GenBank mRNA AY247970.1 - 4, 5 646
BX097338.1 - 1, 2 78

View Full Product Details