Gene Transcripts

Gene Symbol
OR4K17
Entrez Gene ID
390436
Gene Name
olfactory receptor family 4 subfamily K member 17
Gene Aliases
OR14-29
Chromosome Location
Chr. 14: 20117407 - 20118438 on Build GRCh38
UniGene ID
Hs.553765
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004715.1 NP_001004715.1 1 933

View Full Product Details