Gene Transcripts

Gene Symbol
OR11H4
Entrez Gene ID
390442
Gene Name
olfactory receptor family 11 subfamily H member 4
Gene Aliases
OR14-36
Chromosome Location
Chr. 14: 20242792 - 20243766 on Build GRCh38
UniGene ID
Hs.553767
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004479.1 NP_001004479.1 1 834
GenBank mRNA BC137054.1 - 1 887
BC137055.1 - 1 887 9021052

View Full Product Details