Gene Transcripts

Gene Symbol
FGFRL1
Entrez Gene ID
53834
Gene Name
fibroblast growth factor receptor-like 1
Gene Aliases
FGFR5, FHFR
Chromosome Location
Chr. 4: 1011822 - 1026898 on Build GRCh38
UniGene ID
Hs.193326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004356.2 NP_001004356.1 6 1109
NM_001004358.1 NP_001004358.1 6 992
NM_021923.3 NP_068742.2 5 982
XM_011513486.1 XP_011511788.1 6 1113
GenBank mRNA AF279689.1 - 6 992
AF312678.1 - 1109
AJ277437.1 - 5 982
AK172829.1 - 5 966
AK225486.1 - 5 953
AK314365.1 - 6 1110
AY358303.1 - 1109
BC015228.1 - 2 393
BC036769.1 - 6 1121 5744684

View Full Product Details