Gene Transcripts

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
Chromosome Location
Chr. 1: 205504556 - 205532793 on Build GRCh38
UniGene ID
Hs.445402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002596.3 NP_002587.2 4 671
NM_212502.2 NP_997667.1 4 589
NM_212503.2 NP_997668.1 4 679
XM_011509602.2 XP_011507904.1 4 729
XM_017001423.1 XP_016856912.1 4 824
GenBank mRNA AB209307.1 - 2 1251
AK293756.1 - 3 953
AK295272.1 - 2 119
AK295654.1 - 3 734
AK296320.1 - 4 637
AK308913.1 - 2 120
AU130437.1 - 4 550
AY353237.1 - 4 504
AY353238.1 - 4 568
AY353239.1 - 4 391
BC011526.1 - 4 524 3855304
BT007299.1 - 3 321
BU165355.1 - 591
X66362.1 - 1 45

View Full Product Details