Gene Transcripts

Gene Symbol
FCRL2
Entrez Gene ID
79368
Gene Name
Fc receptor like 2
Gene Aliases
CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
Chromosome Location
Chr. 1: 157745733 - 157777472 on Build GRCh38
UniGene ID
Hs.437393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159488.1 NP_001152960.1 6 1121
NM_030764.3 NP_110391.2 6 1121
XM_011509974.2 XP_011508276.1 6 1456
XM_011509975.2 XP_011508277.1 6 1458
XM_011509976.2 XP_011508278.1 6 1457
XM_017002316.1 XP_016857805.1 6 1456
XM_017002319.1 XP_016857808.1 5 1171
GenBank mRNA AF319438.1 - 1 606
AF319439.1 - 1 444
AF319440.1 - 1 594
AF390037.1 - 1 444
AF459633.1 - 6 1109
AK301113.1 - 6 1108
AL833194.1 - 1 2980
AY043465.1 - 6 1108
AY358130.1 - 6 1111
BC129836.1 - 1 446 40115446

View Full Product Details