Gene Transcripts

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
Chromosome Location
Chr. X: 14690863 - 14873255 on Build GRCh38
UniGene ID
Hs.554740
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018113.2 NP_001018123.1 8 1973
NM_001324162.1 NP_001311091.1 8 1952
NM_152633.3 NP_689846.1 7 1852
XM_011545470.2 XP_011543772.1 8 2159
XM_017029355.1 XP_016884844.1 8 2051
XM_017029356.1 XP_016884845.1 8 2159
GenBank mRNA AK091383.1 - 7 1852
BC043596.1 - 8 1956
BC136558.1 - 8 1955 9020548
BC136560.1 - 7 1834 9020550

View Full Product Details