Gene Transcripts
- Gene Symbol
- GLI4
- Entrez Gene ID
- 2738
- Gene Name
- GLI family zinc finger 4
- Gene Aliases
- HKR4, ZNF928
- Chromosome Location
- Chr. 8: 143267437 - 143276931 on Build GRCh38
- UniGene ID
- Hs.400533
- Species
- Homo sapiens
- Gene Symbol
- ZFP41
- Entrez Gene ID
- 286128
- Gene Name
- ZFP41 zinc finger protein
- Gene Aliases
- ZNF753, zfp-41
- Chromosome Location
- Chr. 8: 143246821 - 143262705 on Build GRCh38
- UniGene ID
- Hs.668016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL831951.1 |
- |
4 |
2735 |
|
View Full Product Details