Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001198754.1 | NP_001185683.1 | 3 | 318 | |
NM_001198755.1 | NP_001185684.1 | 3 | 345 | ||
NM_001198756.1 | NP_001185685.1 | 3 | 235 | ||
NM_031498.2 | NP_113686.1 | 3 | 362 | ||
GenBank mRNA | AF365870.1 | - | 2 | 126 | |
AF493876.1 | - | 1 | 55 | ||
AK312839.1 | - | 3 | 346 | ||
BC008663.1 | - | 3 | 362 | 3866242 | |
BM723861.1 | - | 3 | 235 | ||
DA248836.1 | - | 3 | 318 |