Gene Transcripts

Gene Symbol
GPR27
Entrez Gene ID
2850
Gene Name
G protein-coupled receptor 27
Gene Aliases
SREB1
Chromosome Location
Chr. 3: 71754050 - 71755177 on Build GRCh38
UniGene ID
Hs.591653
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018971.1 NP_061844.1 1 1104
GenBank mRNA AB040799.1 - 1 1104
HQ709187.1 - 1 1104

View Full Product Details