Gene Transcripts

Gene Symbol
LIMS1
Entrez Gene ID
3987
Gene Name
LIM zinc finger domain containing 1
Gene Aliases
PINCH, PINCH-1, PINCH1
Chromosome Location
Chr. 2: 108534355 - 108687246 on Build GRCh38
UniGene ID
Hs.597715
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193482.1 NP_001180411.1 6 639
NM_001193483.2 NP_001180412.1 6 784
NM_001193484.1 NP_001180413.1 6 760
NM_001193485.2 NP_001180414.1 6 989
NM_001193488.1 NP_001180417.1 6 707
NM_004987.5 NP_004978.2 6 669
XM_005263946.1 XP_005264003.1 6 789
XM_005263948.3 XP_005264005.1 6 842
XM_005263949.1 XP_005264006.1 6 784
XM_005263950.1 XP_005264007.1 6 625
XM_005263951.4 XP_005264008.1 6 1061
XM_017004091.1 XP_016859580.1 6 811
XM_017004092.1 XP_016859581.1 6 806
XM_017004093.1 XP_016859582.1 6 855
XM_017004094.1 XP_016859583.1 6 1090
GenBank mRNA AF527759.1 - 6 989
AF527760.1 - 6 741
AF527761.1 - 6 717
AF527762.1 - 6 650
AF527763.1 - 6 639
AK296992.1 - 6 739
AK302411.1 - 6 764
AK304260.1 - 6 760
AK314217.1 - 6 669
BC005341.1 - 6 707 3838632
U09284.2 - 6 659

View Full Product Details