Gene Transcripts
- Gene Symbol
- PPP2R3A
- Entrez Gene ID
- 5523
- Gene Name
- protein phosphatase 2 regulatory subunit B''alpha
- Gene Aliases
- PPP2R3, PR130, PR72
- Chromosome Location
- Chr. 3: 135965673 - 136147910 on Build GRCh38
- UniGene ID
- Hs.518155
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190447.1 |
NP_001177376.1
|
5, 6 |
729 |
|
NM_002718.4 |
NP_002709.2
|
6, 7 |
3164 |
|
NM_181897.2 |
NP_871626.1
|
5, 6 |
1019 |
|
XM_006713686.3 |
XP_006713749.1
|
6, 7 |
3575 |
|
XM_011512956.2 |
XP_011511258.1
|
6, 7 |
3339 |
|
XM_017006787.1 |
XP_016862276.1
|
5, 6 |
2897 |
|
GenBank mRNA |
AK293012.1 |
- |
6, 7 |
3144 |
|
AK298059.1 |
- |
5, 6 |
658 |
|
AK299544.1 |
- |
5, 6 |
689 |
|
AK302163.1 |
- |
4, 5 |
427 |
|
AK314243.1 |
- |
6, 7 |
3092 |
|
AK316258.1 |
- |
5, 6 |
972 |
|
AL389975.1 |
- |
3, 4 |
247 |
|
BC065531.1 |
- |
6, 7 |
3111 |
6161260 |
L07590.1 |
- |
6, 7 |
3051 |
|
L12146.1 |
- |
5, 6 |
944 |
|
View Full Product Details