Gene Transcripts

Gene Symbol
PPP2R3A
Entrez Gene ID
5523
Gene Name
protein phosphatase 2 regulatory subunit B''alpha
Gene Aliases
PPP2R3, PR130, PR72
Chromosome Location
Chr. 3: 135965673 - 136147910 on Build GRCh38
UniGene ID
Hs.518155
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190447.1 NP_001177376.1 7 949
NM_002718.4 NP_002709.2 8 3384
NM_181897.2 NP_871626.1 7 1239
XM_006713686.3 XP_006713749.1 8 3795
XM_011512956.2 XP_011511258.1 8 3559
XM_017006787.1 XP_016862276.1 7 3117
GenBank mRNA AK293012.1 - 8 3364
AK298059.1 - 7 878
AK299544.1 - 7 909
AK302163.1 - 7 781
AK314243.1 - 8 3312
AK316258.1 - 7 1192
AL389975.1 - 5 467
BC065531.1 - 8 3331 6161260
L07590.1 - 8 3271
L12146.1 - 7 1164

View Full Product Details