Gene Transcripts

Gene Symbol
CLEC3B
Entrez Gene ID
7123
Gene Name
C-type lectin domain family 3 member B
Gene Aliases
TN, TNA
Chromosome Location
Chr. 3: 45026207 - 45036073 on Build GRCh38
UniGene ID
Hs.476092
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308394.1 NP_001295323.1 3 341
NM_003278.2 NP_003269.2 3 438
XM_017007116.1 XP_016862605.1 2 399
XM_017007117.1 XP_016862606.1 2 964
GenBank mRNA AK312093.1 - 3 446
BC011024.1 - 3 403 4042921
BI826174.1 - 3 341
BI826652.1 - 3 307
BI837909.1 - 3 522
BM669340.1 - 1 428
CD616729.1 - 3 412
CR541981.1 - 3 342
CR542009.1 - 3 342
CR749292.1 - 3 4320
X64559.1 - 3 435

View Full Product Details