Gene Transcripts
- Gene Symbol
- TPD52L2
- Entrez Gene ID
- 7165
- Gene Name
- tumor protein D52 like 2
- Gene Aliases
- D54, TPD54
- Chromosome Location
- Chr. 20: 63865228 - 63891545 on Build GRCh38
- UniGene ID
- Hs.473296
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243891.1 |
NP_001230820.1
|
6, 7 |
578 |
|
NM_001243892.1 |
NP_001230821.1
|
5, 6 |
524 |
|
NM_001243894.1 |
NP_001230823.1
|
4, 5 |
491 |
|
NM_003288.3 |
NP_003279.2
|
6, 7 |
653 |
|
NM_199359.2 |
NP_955391.1
|
5, 6 |
593 |
|
NM_199360.2 |
NP_955392.1
|
8, 9 |
722 |
|
NM_199361.2 |
NP_955393.1
|
7, 8 |
662 |
|
NM_199362.2 |
NP_955394.1
|
7, 8 |
695 |
|
NM_199363.2 |
NP_955395.1
|
6, 7 |
635 |
|
NR_045090.1 |
|
6, 7 |
622 |
|
GenBank mRNA |
AB062396.1 |
- |
2, 3 |
111 |
|
AF004429.1 |
- |
3, 4 |
100 |
|
AF004430.1 |
- |
6, 7 |
591 |
|
AK055068.1 |
- |
6, 7 |
594 |
|
AK128744.1 |
- |
1, 2 |
2347 |
|
AK223031.1 |
- |
6, 7 |
565 |
|
AK293349.1 |
- |
6, 7 |
475 |
|
AK297064.1 |
- |
5, 6 |
551 |
|
AK298366.1 |
- |
5, 6 |
509 |
|
AK314501.1 |
- |
6, 7 |
611 |
|
AU124850.1 |
- |
6, 7 |
582 |
|
AY323819.1 |
- |
6, 7 |
522 |
|
BC006804.1 |
- |
6, 7 |
538 |
3446037 |
BI667351.1 |
- |
7, 8 |
474 |
|
BI754201.1 |
- |
7, 8 |
549 |
|
BM473873.1 |
- |
6, 7 |
610 |
|
BP347669.1 |
- |
|
560 |
|
BQ948007.1 |
- |
6, 7 |
548 |
|
BT019631.1 |
- |
7, 8 |
557 |
|
CR542036.1 |
- |
6, 7 |
515 |
|
CR749208.1 |
- |
5, 6 |
492 |
|
DA689198.1 |
- |
4, 5 |
390 |
|
DC362382.1 |
- |
5, 6 |
509 |
|
DC428780.1 |
- |
6, 7 |
508 |
|
View Full Product Details