Gene Transcripts
- Gene Symbol
- ZNF20
- Entrez Gene ID
- 7568
- Gene Name
- zinc finger protein 20
- Gene Aliases
- KOX13
- Chromosome Location
- Chr. 19: 12131350 - 12140407 on Build GRCh38
- UniGene ID
- Hs.512823
- Species
- Homo sapiens
- Gene Symbol
- ZNF625-ZNF20
- Entrez Gene ID
- 100529855
- Gene Name
- ZNF625-ZNF20 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 19: 12131350 - 12156731 on Build GRCh38
- UniGene ID
- Hs.512823
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037802.1 |
|
8 |
2725 |
|
View Full Product Details