Gene Transcripts
- Gene Symbol
- MMP23B
- Entrez Gene ID
- 8510
- Gene Name
- matrix metallopeptidase 23B
- Gene Aliases
- MIFR, MIFR-1, MMP22, MMP23A
- Chromosome Location
- Chr. 1: 1631681 - 1634654 on Build GRCh38
- UniGene ID
- Hs.192316
- Species
- Homo sapiens
- Gene Symbol
- MMP23A
- Entrez Gene ID
- 8511
- Gene Name
- matrix metallopeptidase 23A (pseudogene)
- Gene Aliases
- MIFR, MIFR-1, MMP21
- Chromosome Location
- Chr. 1: 1699939 - 1701808 on Build GRCh38
- UniGene ID
- Hs.671760
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002946.1 |
|
2 |
250 |
|
GenBank mRNA |
AF057061.1 |
- |
1 |
101 |
|
AF057062.1 |
- |
1 |
101 |
|
View Full Product Details