Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006049.2 | NP_006040.1 | 2 | 244 | |
GenBank mRNA | AF093593.1 | - | 2 | 167 | |
AK312018.1 | - | 2 | 216 | ||
CD685245.1 | - | 2 | 281 | ||
CN347010.1 | - | 2 | 166 | ||
DB037177.1 | - | 2 | 176 |