Gene Transcripts

Gene Symbol
CLEC4M
Entrez Gene ID
10332
Gene Name
C-type lectin domain family 4 member M
Gene Aliases
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
Chromosome Location
Chr. 19: 7763052 - 7770559 on Build GRCh38
UniGene ID
Hs.421437
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144904.1 NP_001138376.1 4 734
NM_001144905.1 NP_001138377.1 5 815
NM_001144909.1 NP_001138381.1 5 749
NM_001144910.1 NP_001138382.1 5 818
NM_001144911.1 NP_001138383.1 3 803
NM_014257.4 NP_055072.3 4 887
NR_026707.1 4 887
NR_026708.1 4 887
NR_026709.1 3 824
XM_006722611.2 XP_006722674.1 4 819
XM_006722612.3 XP_006722675.1 3 893
XM_006722613.3 XP_006722676.1 3 895
XM_006722614.3 XP_006722677.1 2 811
XM_006722615.2 XP_006722678.1 4 887
GenBank mRNA AB015629.1 - 3 709
AF245219.2 - 4 793
AF290887.1 - 5 753
AK023750.1 - 2 608
AK292278.1 - 5 814
AK298084.1 - 3 708
AK304499.1 - 4 667
AK314704.1 - 5 748
AY042234.1 - 4 769
AY042235.1 - 4 769
AY042236.1 - 4 769
AY042239.1 - 3 706
AY343913.1 - 5 632
BC038851.1 - 4 841 5166966
BC110614.2 - 2 742 40032453

View Full Product Details