Gene Transcripts

Gene Symbol
BAIAP2
Entrez Gene ID
10458
Gene Name
BAI1 associated protein 2
Gene Aliases
BAP2, FLAF3, IRSP53
Chromosome Location
Chr. 17: 81035147 - 81117432 on Build GRCh38
UniGene ID
Hs.128316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144888.1 NP_001138360.1 4 367
NM_006340.2 NP_006331.1 4 367
NM_017450.2 NP_059344.1 4 367
NM_017451.2 NP_059345.1 4 367
XM_005256943.1 XP_005257000.1 5 466
XM_005256944.1 XP_005257001.1 5 466
XM_005256945.1 XP_005257002.1 5 466
XM_005256948.3 XP_005257005.1 4 367
XM_006721635.1 XP_006721698.1 5 466
XM_006721636.1 XP_006721699.1 5 466
XM_006721637.2 XP_006721700.1 4 367
XM_011524193.1 XP_011522495.1 5 466
XM_011524194.1 XP_011522496.1 5 466
XM_017024017.1 XP_016879506.1 3 1124
XM_017024018.1 XP_016879507.1 2 586
GenBank mRNA AB015019.1 - 4 352
AB015020.1 - 4 352
AB017119.1 - 4 352
AB017120.1 - 4 352
AK023688.1 - 5 462
AK225612.1 - 4 364
AK298629.1 - 2 202
BC002495.2 - 4 342
BC014020.2 - 4 354 4125501
BC032559.1 - 4 365 5562784
CB153324.1 - 4 392
U70669.1 - 4 352

View Full Product Details