Gene Transcripts
- Gene Symbol
- DENND2A
- Entrez Gene ID
- 27147
- Gene Name
- DENN domain containing 2A
- Gene Aliases
- FAM31D, KIAA1277
- Chromosome Location
- Chr. 7: 140518418 - 140642209 on Build GRCh38
- UniGene ID
- Hs.6385
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318052.1 |
NP_001304981.1
|
10, 11 |
2375 |
|
NM_001318053.1 |
NP_001304982.1
|
11, 12 |
2285 |
|
NM_015689.4 |
NP_056504.3
|
11, 12 |
2477 |
|
NR_134477.1 |
|
9, 10 |
2102 |
|
XM_005249977.3 |
XP_005250034.1
|
11, 12 |
3008 |
|
XM_005249979.4 |
XP_005250036.1
|
11, 12 |
2261 |
|
XM_011516052.2 |
XP_011514354.1
|
11, 12 |
2355 |
|
XM_011516053.2 |
XP_011514355.1
|
10, 11 |
2459 |
|
XM_011516054.2 |
XP_011514356.1
|
11, 12 |
2260 |
|
XM_017011989.1 |
XP_016867478.1
|
10, 11 |
2461 |
|
GenBank mRNA |
AB033103.2 |
- |
10, 11 |
2375 |
|
AK290056.1 |
- |
11, 12 |
2246 |
|
BC049193.1 |
- |
7, 8 |
845 |
|
BC115004.1 |
- |
11, 12 |
2285 |
7470138 |
BC132875.1 |
- |
9, 10 |
2072 |
40146897 |
View Full Product Details