Gene Transcripts

Gene Symbol
MCAT
Entrez Gene ID
27349
Gene Name
malonyl-CoA-acyl carrier protein transacylase
Gene Aliases
FASN2C, MCT, MT, NET62, fabD
Chromosome Location
Chr. 22: 43132206 - 43143397 on Build GRCh38
UniGene ID
Hs.349111
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014507.3 NP_055322.1 3 818
NM_173467.4 NP_775738.3 4 1036
NR_046423.1 3 852
XM_011530156.2 XP_011528458.1 4 650
GenBank mRNA AA402283.1 - 3 382
AL359401.1 - 4 1003
AL359403.1 - 3 785
BC030985.1 - 3 770
BC042195.1 - 4 1029 6052380
CA422238.1 - 1 362

View Full Product Details