Gene Transcripts

Gene Symbol
THSD1
Entrez Gene ID
55901
Gene Name
thrombospondin type 1 domain containing 1
Gene Aliases
TMTSP, UNQ3010
Chromosome Location
Chr. 13: 52377167 - 52406494 on Build GRCh38
UniGene ID
Hs.325667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018676.3 NP_061146.1 3 1260
NM_199263.2 NP_954872.1 3 1260
GenBank mRNA AB044385.1 - 3 866
AK096289.1 - 3 1260
AK315087.1 - 3 909
AY358149.1 - 3 923
BC063842.1 - 3 893 6181100

View Full Product Details