Gene Transcripts
- Gene Symbol
- FAM91A1
- Entrez Gene ID
- 157769
- Gene Name
- family with sequence similarity 91 member A1
- Gene Aliases
- Chromosome Location
- Chr. 8: 123768439 - 123815452 on Build GRCh38
- UniGene ID
- Hs.459174
- Species
- Homo sapiens
- Gene Symbol
- LINC00869
- Entrez Gene ID
- 57234
- Gene Name
- long intergenic non-protein coding RNA 869
- Gene Aliases
- FAM91A2
- Chromosome Location
- Chr. 1: 149607012 - 149679523 on Build GRCh38
- UniGene ID
- Hs.456578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_111950.1 |
|
9 |
3543 |
|
GenBank mRNA |
AB007962.1 |
- |
9 |
3211 |
|
- Gene Symbol
- LINC01138
- Entrez Gene ID
- 388685
- Gene Name
- long intergenic non-protein coding RNA 1138
- Gene Aliases
- LINC00875
- Chromosome Location
- Chr. 1: 148432959 - 148459920 on Build GRCh38
- UniGene ID
- Hs.713362
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027468.3 |
|
2 |
417 |
|
GenBank mRNA |
AK310186.1 |
- |
2 |
325 |
|
View Full Product Details