Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178439.4 | NP_848526.1 | 5 | 898 | |
XM_011533033.1 | XP_011531335.1 | 5 | 862 | ||
XM_011533034.2 | XP_011531336.1 | 5 | 905 | ||
XM_017004704.1 | XP_016860193.1 | 6 | 927 | ||
XM_017004705.1 | XP_016860194.1 | 3 | 172 | ||
GenBank mRNA | AK023119.1 | - | 5 | 804 | |
AK023261.1 | - | 4 | 240 | ||
BC007420.2 | - | 5 | 818 | 3030159 |