Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318736.1 | NP_001305665.1 | 11 | 1961 | |
NM_024725.3 | NP_079001.2 | 10 | 1826 | ||
XM_005274292.3 | XP_005274349.1 | 8 | 1706 | ||
XM_006718911.3 | XP_006718974.1 | 11 | 1778 | ||
XM_011542984.2 | XP_011541286.1 | 10 | 2490 | ||
XM_011542987.2 | XP_011541289.1 | 11 | 1922 | ||
XM_011542988.2 | XP_011541290.1 | 9 | 1798 | ||
XM_017018305.1 | XP_016873794.1 | 9 | 3571 | ||
XM_017018306.1 | XP_016873795.1 | 10 | 3655 | ||
XM_017018307.1 | XP_016873796.1 | 11 | 2297 | ||
XM_017018308.1 | XP_016873797.1 | 8 | 3531 | ||
XM_017018309.1 | XP_016873798.1 | 8 | 6571 | ||
XM_017018310.1 | XP_016873799.1 | 8 | 6612 | ||
XM_017018311.1 | XP_016873800.1 | 10 | 3502 | ||
XM_017018312.1 | XP_016873801.1 | 11 | 2421 | ||
GenBank mRNA | AF245436.1 | - | 10 | 1814 | |
AK027171.1 | - | 7 | 1032 | ||
AK056790.1 | - | 10 | 1819 | ||
AK074306.1 | - | 11 | 1954 | ||
AK314669.1 | - | 7 | 1855 | ||
BC033726.2 | - | 7 | 1627 | 5001231 |